(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3,3,3-Trifluoropropyl)trichlorosilane is used for the production of fluoro silicones liquids.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Omar W. Steward.; Ogden R. Pierce. The chlorination of (3,3,3-trifluoropropyl)trichlorosilane. Journal of Organometallic Chemistry. 1965, 4, (2), 138-144.
- Paul Tarrant.; G. W. Dyckes.; Robert Dunmire.; G. B. Butler. The Preparation of Some Fluoroalkylmethyldichlorosilanes and their Hydrolysis Products. J. Am. Chem. Soc. 1957, 79, (24), 6536-6540.