Thermo Scientific Chemicals

(3,3,3-Trifluoropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: L16671.06
5 g, Each
Thermo Scientific Chemicals

(3,3,3-Trifluoropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: L16671.06
5 g, Each
Quantity
Catalog number: L16671.06
also known as L16671-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
429-60-7
IUPAC Name
trimethoxy(3,3,3-trifluoropropyl)silane
Molecular Formula
C6H13F3O3Si
InChI Key
JLGNHOJUQFHYEZ-UHFFFAOYSA-N
SMILES
CO[Si](CCC(F)(F)F)(OC)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥ 96.0%
Infrared spectrum
Conforms

Description

(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3,3,3-Trifluoropropyl)trimethoxysilane is used in the manufacture of defoaming agents, releasing agents, fluoro silicone resin and coating. It is used as a coupling agent for epoxy resin.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text