Bis(2,2,2-trifluoroethyl)amine, 95%, Thermo Scientific Chemicals
Bis(2,2,2-trifluoroethyl)amine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(2,2,2-trifluoroethyl)amine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L16709.06
also known as L16709-06
Price (USD)/ Each
237.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
237.00
Add to cart
Bis(2,2,2-trifluoroethyl)amine, 95%, Thermo Scientific Chemicals
Catalog numberL16709.06
Price (USD)/ Each
237.00
-
Add to cart
Chemical Identifiers
CAS407-01-2
IUPAC Namebis(2,2,2-trifluoroethyl)amine
Molecular FormulaC4H5F6N
InChI KeyGTJGHXLFPMOKCE-UHFFFAOYSA-N
SMILESFC(F)(F)CNCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.2985-1.3045 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only