Thermo Scientific Chemicals

3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: L16740.03
1 g, Each
Thermo Scientific Chemicals

3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: L16740.03
1 g, Each
Quantity
Catalog number: L16740.03
also known as L16740-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
396-64-5
IUPAC Name
3,3'-difluoro-1,1'-biphenyl
Molecular Formula
C12H8F2
InChI Key
GAYJHUJLHJWCTH-UHFFFAOYSA-N
SMILES
FC1=CC(=CC=C1)C1=CC(F)=CC=C1
Appearance (Color)
Clear pale yellow to yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5660-1.5710 @ 20?C

Description

3,3'-Difluorobiphenyl is used materials in ferroelectric display devices

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Difluorobiphenyl is used materials in ferroelectric display devices

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text