3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals
3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L16740.06
also known as L16740-06
Price (USD)/ Each
429.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
429.00
Add to cart
3,3'-Difluorobiphenyl, 97%, Thermo Scientific Chemicals
Catalog numberL16740.06
Price (USD)/ Each
429.00
-
Add to cart
Chemical Identifiers
CAS396-64-5
IUPAC Name3,3'-difluoro-1,1'-biphenyl
Molecular FormulaC12H8F2
InChI KeyGAYJHUJLHJWCTH-UHFFFAOYSA-N
SMILESFC1=CC(=CC=C1)C1=CC(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5660-1.5710 @ 20?C
3,3'-Difluorobiphenyl is used materials in ferroelectric display devices

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Difluorobiphenyl is used materials in ferroelectric display devices

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Jacob H. Goldstein.; A. R. Tarpley. Nuclear magnetic resonance spectra and substituent effects for symmetrically substituted dihalobiphenyls. J. Phys. Chem. 1971, 73, (3), 421-430.
  2. A.L. Allred.; L.W. Bush. Anion radicals of some fluorobiphenyls. Tetrahedron. 1968 , 24, (24), 6883-6887.