Thermo Scientific Chemicals

Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals

Catalog number: L16757.14
25 g, Each
Thermo Scientific Chemicals

Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals

Catalog number: L16757.14
25 g, Each
Quantity
Catalog number: L16757.14
also known as L16757-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2994-71-0
Form
Liquid
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Refractive Index
≤1.3000 @ 20?C

Description

Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text