Thermo Scientific Chemicals

3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals

Catalog number: L16775.MF
50mg, Each
Thermo Scientific Chemicals

3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals

Catalog number: L16775.MF
50mg, Each
Quantity
Catalog number: L16775.MF
also known as L16775-MF
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7423-96-3
IUPAC Name
(2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
Molecular Formula
C9H10FNO3
InChI Key
VIIAUOZUUGXERI-JLDDOWRYNA-N
SMILES
N[C@@H](CC1=CC=C(O)C(F)=C1)C(O)=O
Appearance (Color)
White to cream or pale brown
Form
Powder
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Optical Rotation
-6.0 to -10.0? (c=1 in 1m HCl)

Description

3-Fluoro-L-tyrosine is used as pharma and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-L-tyrosine is used as pharma and intermediates.

Solubility
Soluble in water (9.8 g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text