Nonafluoro-2-iodobutane, 98%, stab. with copper, Thermo Scientific Chemicals
Nonafluoro-2-iodobutane, 98%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nonafluoro-2-iodobutane, 98%, stab. with copper, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16792.06
also known as L16792-06
Price (USD)/ Each
53.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
53.70
Add to cart
Nonafluoro-2-iodobutane, 98%, stab. with copper, Thermo Scientific Chemicals
Catalog numberL16792.06
Price (USD)/ Each
53.70
-
Add to cart
Chemical Identifiers
CAS375-51-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.3290-1.3330 @ 20°C
Appearance (Color)Clear colorless to pale yellow or pale pink or purple or blue-green
StabilizerCopper powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only