1H,1H,10H,10H-Perfluoro-1,10-decanediol, 96%, Thermo Scientific Chemicals
1H,1H,10H,10H-Perfluoro-1,10-decanediol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1H,1H,10H,10H-Perfluoro-1,10-decanediol, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L16823.06
also known as L16823-06
Price (USD)/ Each
78.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
78.10
Add to cart
1H,1H,10H,10H-Perfluoro-1,10-decanediol, 96%, Thermo Scientific Chemicals
Catalog numberL16823.06
Price (USD)/ Each
78.10
-
Add to cart
Chemical Identifiers
CAS754-96-1
IUPAC Name2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorodecane-1,10-diol
Molecular FormulaC10H6F16O2
InChI KeyNSKCTPBWPZPFHW-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)>95.0%
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only