Thermo Scientific Chemicals

3,3,3-Trifluoro-1-propanol, 97%, Thermo Scientific Chemicals

Catalog number: L16879.06
5 g, Each
Thermo Scientific Chemicals

3,3,3-Trifluoro-1-propanol, 97%, Thermo Scientific Chemicals

Catalog number: L16879.06
5 g, Each
Quantity
Catalog number: L16879.06
also known as L16879-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2240-88-2
IUPAC Name
3,3,3-trifluoropropan-1-ol
Molecular Formula
C3H5F3O
InChI Key
HDBGBTNNPRCVND-UHFFFAOYSA-N
SMILES
OCCC(F)(F)F
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.3215-1.3265 @ 20?C

Description

3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-1-propanol is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text