2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine, Mass Spec Std., Thermo Scientific Chemicals
Catalog numberL16883.06
Price (USD)/ Each
174.00
-
Add to cart
Specifications
Chemical Name or Material2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
Name NoteMass Spec Standard
CAS915-76-4
Recommended StorageAmbient temperatures
Density1.716
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.