Thermo Scientific Chemicals

Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: L16891.03
1 g, Each
Thermo Scientific Chemicals

Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%, Thermo Scientific Chemicals

Catalog number: L16891.03
1 g, Each
Quantity
Catalog number: L16891.03
also known as L16891-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28523-86-6
IUPAC Name
1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
Molecular Formula
C4H3F7O
InChI Key
DFEYYRMXOJXZRJ-UHFFFAOYSA-N
SMILES
FCOC(C(F)(F)F)C(F)(F)F
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
<1.3000 @ 20?C

Description

Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text