Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%, Thermo Scientific Chemicals
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L16891.06
also known as L16891-06
Price (USD)/ Each
252.65
Online exclusive
280.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
252.65
Online exclusive
280.00 
Save 27.35 (10%)
Add to cart
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, 98+%, Thermo Scientific Chemicals
Catalog numberL16891.06
Price (USD)/ Each
252.65
Online exclusive
280.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS28523-86-6
IUPAC Name1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
Molecular FormulaC4H3F7O
InChI KeyDFEYYRMXOJXZRJ-UHFFFAOYSA-N
SMILESFCOC(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index<1.3000 @ 20?C
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether is a volatile anesthetic, which inhibits the activation of inflammatory neutrophil and granulocyte in human blood during simulated extracorporeal circulation. It also serves as an antibacterial and used as an inhalational anaesthetic for induction and maintenance of general anesthesia. Further, it finds application in anesthesia of children and infants to make them asleep for surgery. In addition to this, it is used in veterinary medicine.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kawada, C.; Kimura, M.; Masuda, Y.; Nomura, Y. Oral administration of hyaluronan prevents skin dryness and epidermal thickening in ultraviolet irradiated hairless mice. J. Photochem. Photobiol., B 2015, 153, 215-221.
  2. Munekage, M.; Yatabe, T.; Kitagawa, H.; Takezaki, Y.; Tamura, T.; Namikawa, T.; Hanazaki, K. An artificial pancreas provided a novel model of blood glucose level variability in beagles. J. Artif. Organs 2015, 18 (4), 387-390.