Thermo Scientific Chemicals

1,3,5-Triazine, 97%, Thermo Scientific Chemicals

Catalog number: L16911.03
1 g, Each
Thermo Scientific Chemicals

1,3,5-Triazine, 97%, Thermo Scientific Chemicals

Catalog number: L16911.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L16911.03
also known as L16911-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
290-87-9
IUPAC Name
1,3,5-triazine
Molecular Formula
C3H3N3
InChI Key
JIHQDMXYYFUGFV-UHFFFAOYSA-N
SMILES
C1=NC=NC=N1
Assay (GC)
≥96.0%
Appearance (Color)
White to cream to yellow
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.5%
Melting Point (clear melt)
72.0-83.0?C

Description

1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

Solubility
Soluble in ethanol (50 mg/ml), and methanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text