3H,5H-Octamethyltetrasiloxane, 96%, Thermo Scientific Chemicals
3H,5H-Octamethyltetrasiloxane, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3H,5H-Octamethyltetrasiloxane, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16956.06
also known as L16956-06
Price (USD)/ Each
43.30
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
43.30
Add to cart
3H,5H-Octamethyltetrasiloxane, 96%, Thermo Scientific Chemicals
Catalog numberL16956.06
Price (USD)/ Each
43.30
-
Add to cart
Chemical Identifiers
CAS16066-09-4
Molecular FormulaC8H24O3Si4
InChI KeyOHSYWAVRSCQMHG-UHFFFAOYSA-N
SMILESC[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C
Molecular Weight (g/mol)280.62
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 95.0%
Appearance (Color)Clear, colourless
Refractive Index1.3830 - 1.3890 @20?C
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only