5H-Perfluoropentanoic acid, 96%, Thermo Scientific Chemicals
5H-Perfluoropentanoic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5H-Perfluoropentanoic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L16960.06
also known as L16960-06
Price (USD)/ Each
44.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
44.90
Add to cart
5H-Perfluoropentanoic acid, 96%, Thermo Scientific Chemicals
Catalog numberL16960.06
Price (USD)/ Each
44.90
-
Add to cart
Chemical Identifiers
CAS376-72-7
IUPAC Name2,2,3,3,4,4,5,5-octafluoropentanoic acid
Molecular FormulaC5H2F8O2
InChI KeyVGFKXVSMDOKOJZ-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (GC)≥95.0%
Refractive Index1.3150-1.3210 @ 20?C
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only