Thermo Scientific Chemicals

2-Chloro-2,2-difluoroacetophenone, 96%, Thermo Scientific Chemicals

Catalog number: L16963.03
1 g, Each
Thermo Scientific Chemicals

2-Chloro-2,2-difluoroacetophenone, 96%, Thermo Scientific Chemicals

Catalog number: L16963.03
1 g, Each
Quantity
Catalog number: L16963.03
also known as L16963-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
384-67-8
IUPAC Name
2-chloro-2,2-difluoro-1-phenylethan-1-one
Molecular Formula
C8H5ClF2O
InChI Key
MNOONJNILVDLSW-UHFFFAOYSA-N
SMILES
FC(F)(Cl)C(=O)C1=CC=CC=C1
Refractive Index
1.4925-1.4985 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥95.0%

Description

Reagent is an effective product for the synthesis of difluoromethylated phenols in the presence of mild base and aqueous solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent is an effective product for the synthesis of difluoromethylated phenols in the presence of mild base and aqueous solvent.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text