Thermo Scientific Chemicals

n-Decyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L16965.14
25 g, Each
Thermo Scientific Chemicals

n-Decyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: L16965.14
25 g, Each
Quantity
Catalog number: L16965.14
also known as L16965-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2943-73-9
IUPAC Name
decyltriethoxysilane
Molecular Formula
C16H36O3Si
InChI Key
BAAAEEDPKUHLID-UHFFFAOYSA-N
SMILES
CCCCCCCCCC[Si](OCC)(OCC)OCC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4205-1.4245 @ 20?C

Description

n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text