Thermo Scientific Chemicals

Silicon(IV) oxide, powder, 0.5 micron, 99.9%, Thermo Scientific Chemicals

Catalog number: L16985.14
25 g, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, powder, 0.5 micron, 99.9%, Thermo Scientific Chemicals

Catalog number: L16985.14
25 g, Each
Quantity
Catalog number: L16985.14
also known as L16985-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Color)
White
Form
Powder
Assay (unspecified)
>99.85%
Particle Size
0.4-0.6 micron

Description

Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

Solubility
Insoluble in water.

Notes
It is a very stable compound of silicon.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text