Thermo Scientific Chemicals

5-Bromo-7-nitroindoline, 98%, Thermo Scientific Chemicals

Catalog number: L16993.03
1 g, Each
Thermo Scientific Chemicals

5-Bromo-7-nitroindoline, 98%, Thermo Scientific Chemicals

Catalog number: L16993.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L16993.03
also known as L16993-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
80166-90-1
IUPAC Name
5-bromo-7-nitro-2,3-dihydro-1H-indole
Molecular Formula
C8H7BrN2O2
InChI Key
VXKXMHDXFLFIFI-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC(Br)=CC2=C1NCC2
Appearance (Color)
Orange to dark brown to brown
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%

Description

5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text