Thermo Scientific Chemicals

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 99%, Thermo Scientific Chemicals

Catalog number: L17022.14
25 g, Each
Thermo Scientific Chemicals

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 99%, Thermo Scientific Chemicals

Catalog number: L17022.14
25 g, Each
Quantity
Catalog number: L17022.14
also known as L17022-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23357-52-0
IUPAC Name
1,2,3,4-tetrahydronaphthalen-1-amine
Molecular Formula
C10H13N
InChI Key
JRZGPXSSNPTNMA-UHFFFAOYNA-N
SMILES
NC1CCCC2=CC=CC=C12
Refractive Index
1.5635-1.5655 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥99.0%
Enantiomeric excess
≥99.0%

Description

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text