Thermo Scientific Chemicals

3-Methoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L17050.03
1 g, Each
Thermo Scientific Chemicals

3-Methoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L17050.03
1 g, Each
Quantity
Catalog number: L17050.03
also known as L17050-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10130-74-2
IUPAC Name
3-methoxybenzene-1-sulfonyl chloride
Molecular Formula
C7H7ClO3S
InChI Key
JHJKSEKUZNJKGO-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1)S(Cl)(=O)=O
Appearance (Color)
Clear colorless to yellow or pale purple
Form
Liquid
Assay (GC)
≥96.0% (UK-sourced material)
Assay from Supplier's CofA
≥96.0% (US-sourced material)
Comment
Material sourced in UK and US

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text