Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals
Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L17055.06
also known as L17055-06
Price (USD)/ Each
27.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
27.90
Add to cart
Bis(4-chlorobutyl) ether, 99%, Thermo Scientific Chemicals
Catalog numberL17055.06
Price (USD)/ Each
27.90
-
Add to cart
Chemical Identifiers
CAS6334-96-9
IUPAC Name1-chloro-4-(4-chlorobutoxy)butane
Molecular FormulaC8H16Cl2O
InChI KeyPVBMXMKIKMJQRK-UHFFFAOYSA-N
SMILESClCCCCOCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4570-1.4600 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only