Glycine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Glycine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17116.06
also known as L17116-06
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
Add to cart
Glycine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals
Catalog numberL17116.06
Price (USD)/ Each
153.65
Online exclusive
171.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS27532-96-3
IUPAC Name2-(tert-butoxy)-2-oxoethan-1-aminium
Molecular FormulaC6H14NO2
InChI KeySJMDMGHPMLKLHQ-UHFFFAOYSA-O
SMILESCC(C)(C)OC(=O)C[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Freebase GC)≥96.0%
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

Solubility
Soluble in methanol and water

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. H. R. Allcock.; T. J. Fuller.; D. P. Mack.; K. Matsumura.; Karen M. Smeltz. Synthesis of Poly[(amino acid alkyl ester)phosphazenes]. Macromolecules. 1977, 10 (4), 824-830.
  2. Hiroyuki Ishiwata.; Hiroki Sone.; Hideo Kigoshi.; Kiyoyuki Yamada. Total Synthesis of Doliculide, a Potent Cytotoxic Cyclodepsipeptide from the Japanese Sea Hare Dolabella auricularia. J. Org. Chem. 1994, 59 (17), 4712-4713.