6-Cyanonicotinic acid, 97%, Thermo Scientific Chemicals
6-Cyanonicotinic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Cyanonicotinic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number L17127.03
Price (USD)/ Each
253.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
253.00
Add to cart
6-Cyanonicotinic acid, 97%, Thermo Scientific Chemicals
Catalog numberL17127.03
Price (USD)/ Each
253.00
-
Add to cart
Chemical Identifiers
CAS70165-31-0
IUPAC Name6-cyanopyridine-3-carboxylic acid
Molecular FormulaC7H4N2O2
InChI KeyWMHSQCDPPJRWIL-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to orange or brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Melting Point (clear melt)187-195°C
Assay (HPLC)≥96.0%
6-Cyanonicotinic acid, is used in medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Cyanonicotinic acid, is used in medicine

Solubility
Soluble in alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Rainer Schobert.; Bernhard Biersack. cis-Dichloroplatinum (II) complexes with aminomethylnicotinate and -isonicotinate ligands. Inorganica Chimica Acta. 2005, 358 (12), 3369-3376.