Methyl 3,5-difluorobenzoate, 98%, Thermo Scientific Chemicals
Methyl 3,5-difluorobenzoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3,5-difluorobenzoate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17171.06
also known as L17171-06
Price (USD)/ Each
70.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
70.60
Add to cart
Methyl 3,5-difluorobenzoate, 98%, Thermo Scientific Chemicals
Catalog numberL17171.06
Price (USD)/ Each
70.60
-
Add to cart
Chemical Identifiers
CAS216393-55-4
IUPAC Namemethyl 3,5-difluorobenzoate
Molecular FormulaC8H6F2O2
InChI KeyJBEJPGWPXIIQBB-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(F)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4720-1.4760 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only