Thermo Scientific Chemicals

(R)-(+)-2-Benzyloxypropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: L17205.06
5 g, Each
Thermo Scientific Chemicals

(R)-(+)-2-Benzyloxypropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: L17205.06
5 g, Each
Quantity
Catalog number: L17205.06
also known as L17205-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100836-85-9
IUPAC Name
2-(benzyloxy)propanoic acid
Molecular Formula
C10H12O3
InChI Key
XWAVPOFYNPXXEL-UHFFFAOYNA-N
SMILES
CC(OCC1=CC=CC=C1)C(O)=O
Optical Rotation
+77± 3?, c = 1% in Methanol
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥97.5%

Description

(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Benzyloxypropionic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text