Thermo Scientific Chemicals

1-Ethynylpyrene, 96%, Thermo Scientific Chemicals

Catalog number: L17227.03
1 g, Each
Thermo Scientific Chemicals

1-Ethynylpyrene, 96%, Thermo Scientific Chemicals

Catalog number: L17227.03
1 g, Each
Quantity
Catalog number: L17227.03
also known as L17227-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34993-56-1
IUPAC Name
1-ethynylpyrene
Molecular Formula
C18H10
InChI Key
VEBUBSLYGRMOSZ-UHFFFAOYSA-N
SMILES
C#CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Appearance (Color)
Dark yellow to yellow to orange to brown
Assay (GC)
≥95.0%
Melting Point (clear melt)
109.0-119.0?C
Form
Crystals or powder or crystalline powder or lumps
Assay (HPLC)
≥95.0%

Description

1-Ethynylpyrene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethynylpyrene is used as a pharmaceutical intermediate.

Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text