1-Ethynylpyrene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethynylpyrene is used as a pharmaceutical intermediate.
Solubility
Soluble in chloroform, dichloromethane and toluene. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
Clemens Wagner.; Manuela Rist.; Elke Mayer-Enthart.; Hans-Achim Wagenknecht. 1-Ethynylpyrene-modified guanine and cytosine as optical labels for DNA hybridization. Org. Biomol. Chem. 2005, 3 2062-2063.
Janez Barbarica.; Hans-Achim Wagenknecht. DNA as a supramolecular scaffold for the helical arrangement of a stack of 1-ethynylpyrene chromophores. Org. Biomol. Chem. 2006, 4 (11), 2088-2090.