Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals
Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium hydrogen fluoride, 95%, remainder mainly ammonium fluoride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L17237.0E
Price (USD)/ Each
-
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS1341-49-7
IUPAC Nameamine dihydrofluoride
Molecular FormulaF2H5N
InChI KeyKVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILESN.F.F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>94.0 to <106.0%
FormCrystalline chunks, Flakes or Powder
Appearance (Color)White/translucent
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in a new hydrothermal synthesis of complex fluorides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. L. Erdey.; S. Gal.; G. Liptay. Thermoanalytical properties of analytical-grade reagents: Ammonium salts. Talanta. 1964, 11 (6), 913-940.
  2. B.N. Wani.; U.R.K. Rao. Interfacial reaction between ammonium hydrogen fluoride and binary oxides at room temperature. Materials Chemistry and Physics. 1993, 33 (1-2), 165-167.