Thermo Scientific Chemicals

3-Bromo-N,N-dimethylaniline, 97%, Thermo Scientific Chemicals

Catalog number: L17279.14
25 g, Each
Thermo Scientific Chemicals

3-Bromo-N,N-dimethylaniline, 97%, Thermo Scientific Chemicals

Catalog number: L17279.14
25 g, Each
Quantity
Catalog number: L17279.14
also known as L17279-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16518-62-0
IUPAC Name
3-bromo-N,N-dimethylaniline
Molecular Formula
C8H10BrN
InChI Key
USEXQPWLCGBYNT-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=CC(Br)=C1
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Refractive Index
1.5990-1.6040 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange

Description

3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-N,N-dimethylaniline is involved in fluorination using metal fluoride in the presence of 2-(di-1-adamantylphosphino)-3,6-dimethoxy-2′,4′,6′-tri-i-propyl-1,1′-biphenyl (AdBrettPhos)-based Pd precatalyst, used to prepare 3-fluoro-N,N- dimethylaniline.

Solubility
Miscible with chloroform, dichloromethane and methanol.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text