3-(Trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
3-(Trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17280.06
also known as L17280-06
Price (USD)/ Each
408.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
408.00
Add to cart
3-(Trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Catalog numberL17280.06
Price (USD)/ Each
408.00
-
Add to cart
Chemical Identifiers
CAS3796-23-4
IUPAC Name3-(trifluoromethyl)pyridine
Molecular FormulaC6H4F3N
InChI KeyJTZSFNHHVULOGJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4150-1.4200 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only