Thermo Scientific Chemicals

2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine, 98%, Thermo Scientific Chemicals

Catalog number: L17433.06
5 g, Each
Thermo Scientific Chemicals

2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine, 98%, Thermo Scientific Chemicals

Catalog number: L17433.06
5 g, Each
Quantity
Catalog number: L17433.06
also known as L17433-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
145100-50-1
IUPAC Name
1,1,1-trifluoro-N-(pyridin-2-yl)-N-trifluoromethanesulfonylmethanesulfonamide
Molecular Formula
C7H4F6N2O4S2
InChI Key
DXLQEJHUQKKSRB-UHFFFAOYSA-N
SMILES
FC(F)(F)S(=O)(=O)N(C1=CC=CC=N1)S(=O)(=O)C(F)(F)F
Melting Point (clear melt)
36.0-43.0?C
Appearance (Color)
White to cream to pale yellow or pink
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%

Description

2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine is a reactive agent used for conversion of lithium enolates of ketones to vinyl triflates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[N,N-Bis(trifluoromethylsulfonyl)amino]pyridine is a reactive agent used for conversion of lithium enolates of ketones to vinyl triflates.

Solubility
Hydrolyzes in water.

Notes
It is sensitive to moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text