Thermo Scientific Chemicals

Diphenyliodonium trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: L17444.03
1 g, Each
Thermo Scientific Chemicals

Diphenyliodonium trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: L17444.03
1 g, Each
Quantity
Catalog number: L17444.03
also known as L17444-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66003-76-7
IUPAC Name
diphenyliodanium trifluoromethanesulfonate
Molecular Formula
C13H10F3IO3S
InChI Key
SBQIJPBUMNWUKN-UHFFFAOYSA-M
SMILES
[O-]S(=O)(=O)C(F)(F)F.[I+](C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (unspecified)
≥98.5%
Comment
may contain up to 1.5% excess triflic acid
Elemental Analysis
Carbon, C: 35.74-36.84% (Theory: 36.30%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text