Thermo Scientific Chemicals

1,3-Diiodobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: L17445.06
5 g, Each
Thermo Scientific Chemicals

1,3-Diiodobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: L17445.06
5 g, Each
Quantity
Catalog number: L17445.06
also known as L17445-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-00-6
IUPAC Name
1,3-diiodobenzene
Molecular Formula
C6H4I2
InChI Key
SFPQFQUXAJOWNF-UHFFFAOYSA-N
SMILES
IC1=CC(I)=CC=C1
Appearance (Color)
White to cream to yellow to brown
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
32.0-38.0?C

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in hot methanol.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text