Thermo Scientific Chemicals

4-Iodobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L17448.MD
250 mg, Each
Thermo Scientific Chemicals

4-Iodobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: L17448.MD
250 mg, Each
Quantity
Catalog number: L17448.MD
also known as L17448-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15164-44-0
IUPAC Name
4-iodobenzaldehyde
Molecular Formula
C7H5IO
InChI Key
NIEBHDXUIJSHSL-UHFFFAOYSA-N
SMILES
IC1=CC=C(C=O)C=C1
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to cream to yellow to brown
Assay (GC)
≥98.0%
Free acid (titration)
≤1%

Description

4-Iodobenzaldehyde is used in synthesis of 4-[2-(trimethylsilyl)ethynyl]benzaldehyde, 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin, and 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N?-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzaldehyde is used in synthesis of 4-[2-(trimethylsilyl)ethynyl]benzaldehyde, 5,15-dimesityl-10-(3-[2-(trimethylsilyl)ethynyi]phenyl}-20-(4-iodophenyl)porphyrin, and 5,15-dimesityl-10-[3,5-bis{2-[4-(N,N′-difluoroboryl-1,9-dimethyidipyrrin-5-yl)-phenyl]ethynyl}phenyl]-20-(4-iodophenyl)porphyrin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air and light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text