Thermo Scientific Chemicals

4-Iodobenzylamine, 97%, Thermo Scientific Chemicals

Catalog number: L17450.06
5 g, Each
Thermo Scientific Chemicals

4-Iodobenzylamine, 97%, Thermo Scientific Chemicals

Catalog number: L17450.06
5 g, Each
Quantity
Catalog number: L17450.06
also known as L17450-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39959-59-6
IUPAC Name
1-(4-iodophenyl)methanamine
Molecular Formula
C7H8IN
InChI Key
KCGZGJOBKAXVSU-UHFFFAOYSA-N
SMILES
NCC1=CC=C(I)C=C1
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%
Assay (GC)
≥96.0%
Melting Point (clear melt)
41.0-51.0?C

Description

4-Iodobenzylamine is used to prepare 2-bromo-N-(4-iodo-benzyl)-acetamide in the presence of bromoacetyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzylamine is used to prepare 2-bromo-N-(4-iodo-benzyl)-acetamide in the presence of bromoacetyl bromide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air and light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text