Thermo Scientific Chemicals

4-Bromobenzeneboronic acid neopentyl glycol ester, 98+%, Thermo Scientific Chemicals

Catalog number: L17455.06
5 g, Each
Thermo Scientific Chemicals

4-Bromobenzeneboronic acid neopentyl glycol ester, 98+%, Thermo Scientific Chemicals

Catalog number: L17455.06
5 g, Each
Quantity
Catalog number: L17455.06
also known as L17455-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
183677-71-6
IUPAC Name
2-(4-bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane
Molecular Formula
C11H14BBrO2
InChI Key
GUASPWAWPYERCR-UHFFFAOYSA-N
SMILES
CC1(C)COB(OC1)C1=CC=C(Br)C=C1
Assay (GC)
≥98.0%
Melting Point
107.0-113.0?C
Appearance (Color)
White to pale yellow
Form
Crystals or powder or crystalline powder or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text