1-Boc-pyrrolidine, 98%, Thermo Scientific Chemicals
1-Boc-pyrrolidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-pyrrolidine, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L17475.06
also known as L17475-06
Price (USD)/ Each
68.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
68.70
Add to cart
1-Boc-pyrrolidine, 98%, Thermo Scientific Chemicals
Catalog numberL17475.06
Price (USD)/ Each
68.70
-
Add to cart
Chemical Identifiers
CAS86953-79-9
IUPAC Nametert-butyl pyrrolidine-1-carboxylate
Molecular FormulaC9H17NO2
InChI KeyLPQZERIRKRYGGM-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4465-1.4505 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only