4-Nitrophenyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
4-Nitrophenyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitrophenyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17481.06
also known as L17481-06
Price (USD)/ Each
75.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
75.40
Add to cart
4-Nitrophenyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Catalog numberL17481.06
Price (USD)/ Each
75.40
-
Add to cart
Chemical Identifiers
CAS17763-80-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)50.5-56.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only