4-Bromo-3-methylphenol, 98%, Thermo Scientific Chemicals
4-Bromo-3-methylphenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromo-3-methylphenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L17510.06
also known as L17510-06
Price (USD)/ Each
87.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
87.10
Add to cart
4-Bromo-3-methylphenol, 98%, Thermo Scientific Chemicals
Catalog numberL17510.06
Price (USD)/ Each
87.10
-
Add to cart
Chemical Identifiers
CAS14472-14-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)55.0-62.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only