Thermo Scientific Chemicals

2-Amino-5-chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: L17515.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-5-chlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: L17515.06
5 g, Each
Quantity
Catalog number: L17515.06
also known as L17515-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20028-53-9
IUPAC Name
2-amino-5-chlorobenzaldehyde
Molecular Formula
C7H6ClNO
InChI Key
KUIFMGITZFDQMP-UHFFFAOYSA-N
SMILES
NC1=C(C=O)C=C(Cl)C=C1
Appearance (Color)
Dark yellow to yellow to orange to brown
Form
Crystals or powder or crystalline powder or fused solid or granules
Assay (HPLC)
≥96.0%
Free acid (titration)
≤1.5%
Melting Point (clear melt)
67.0-76.0?C

Description

2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-Chlorobenzaldehyde is used as a raw material in organic synthesis. It also serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text