Thermo Scientific Chemicals

7-Methylindole, 98%, Thermo Scientific Chemicals

Catalog number: L17521.06
5 g, Each
Thermo Scientific Chemicals

7-Methylindole, 98%, Thermo Scientific Chemicals

Catalog number: L17521.06
5 g, Each
Quantity
Catalog number: L17521.06
also known as L17521-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
933-67-5
IUPAC Name
7-methyl-1H-indole
Molecular Formula
C9H9N
InChI Key
KGWPHCDTOLQQEP-UHFFFAOYSA-N
SMILES
CC1=C2NC=CC2=CC=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
78.0-84.0?C
Appearance (Color)
White to cream to yellow to pale brown
Assay (GC)
≥97.5%

Description

It is a reactant for preparation of pharmaceutically active pyrrolidine analogues via Markovnikov addition, tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, plant growth inhibitors, antifungal agents, sodium-Dependent Glucose Co-transporter 2 (SGLT2) Inhibitors for the Management of Hyperglycemia in Diabetes and inhibitors of HIV-1 attachment. It is a reactant for Friedel-Crafts alkylation reactions and reductive formylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for preparation of pharmaceutically active pyrrolidine analogues via Markovnikov addition, tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, plant growth inhibitors, antifungal agents, sodium-Dependent Glucose Co-transporter 2 (SGLT2) Inhibitors for the Management of Hyperglycemia in Diabetes and inhibitors of HIV-1 attachment. It is a reactant for Friedel-Crafts alkylation reactions and reductive formylation reactions.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text