Thermo Scientific Chemicals

1-Boc-isonipecotic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L17527.03
1 g, Each
Thermo Scientific Chemicals

1-Boc-isonipecotic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L17527.03
1 g, Each
Quantity
Catalog number: L17527.03
also known as L17527-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84358-13-4
IUPAC Name
1-[(tert-butoxy)carbonyl]piperidine-4-carboxylic acid
Molecular Formula
C11H19NO4
InChI Key
JWOHBPPVVDQMKB-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCC(CC1)C(O)=O
Assay (Silylated GC)
≥98.0%
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
148.0-154.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
White

Description

It is a pharmaceutical intermediate. he camptothecin ester of isonipecotic acid is installed on a triazine dendrimer intermediate obtained through an iterative, scalable route to ultimately yield cationic and PEGylated targets with activities in cell culture comparable to free drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate. he camptothecin ester of isonipecotic acid is installed on a triazine dendrimer intermediate obtained through an iterative, scalable route to ultimately yield cationic and PEGylated targets with activities in cell culture comparable to free drug.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text