N-(5-Bromopentyl)phthalimide, 97%, Thermo Scientific Chemicals
N-(5-Bromopentyl)phthalimide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(5-Bromopentyl)phthalimide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L17543.06
also known as L17543-06
Price (USD)/ Each
48.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
48.40
Add to cart
N-(5-Bromopentyl)phthalimide, 97%, Thermo Scientific Chemicals
Catalog numberL17543.06
Price (USD)/ Each
48.40
-
Add to cart
Chemical Identifiers
CAS954-81-4
IUPAC Name2-(5-bromopentyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC13H14BrNO2
InChI KeyQKVHAKICMNABGB-UHFFFAOYSA-N
SMILESBrCCCCCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals
Appearance (Color)White to cream or pale yellow
Melting Point (clear melt)54-64?C
Assay (GC)≥96.0%
It is a pharmaceutical intermediate and is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Richard A. Glennon.; Noreen A. Naiman.; Robert A. Lyon.; Milt Titeler. Arylpiperazine derivatives as high-affinity 5-HT1A serotonin ligands.J. Med. Chem. 1988, 31 (10),1968-1971 .
  2. Richard A. Glennon.; Noreen A. Naiman.; M. Edward Pierson.; J. Doyle Smith.; Abd M. Ismaiel.; Milt Titeler.; Robert A. Lyon. N-(phthalimidoalkyl) derivatives of serotonergic agents: a common interaction at 5-HT1A serotonin binding sites?J. Med. Chem. 1989, 32 (8),1921-1926 .