Thermo Scientific Chemicals

N-(5-Bromopentyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: L17543.14
25 g, Each
Thermo Scientific Chemicals

N-(5-Bromopentyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: L17543.14
25 g, Each
Quantity
Catalog number: L17543.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
954-81-4
IUPAC Name
2-(5-bromopentyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C13H14BrNO2
InChI Key
QKVHAKICMNABGB-UHFFFAOYSA-N
SMILES
BrCCCCCN1C(=O)C2=CC=CC=C2C1=O
Form
Powder or crystals
Appearance (Color)
White to cream or pale yellow
Melting Point (clear melt)
54-64?C
Assay (GC)
≥96.0%

Description

It is a pharmaceutical intermediate and is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text