Thermo Scientific Chemicals

Fomblin YR-1800, Thermo Scientific Chemicals

Catalog number: L17548.06
5 g, Each
Thermo Scientific Chemicals

Fomblin YR-1800, Thermo Scientific Chemicals

Catalog number: L17548.06
5 g, Each
Quantity
Catalog number: L17548.06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
69991-67-9
Viscosity
490 - 530 cSt @ 40?C
Appearance (Color)
Clear, colorless
Comment
Fomblin is a registered trademark of Solvay Solexis S.P.A.
Evaporation loss
<0.50% (22hr, 204?C)
Identification (FTIR)
Conforms

Description

It is used in applications include pump and compressor cleaning, degreasing, dewatering, vapor blanketing, flash point reduction, and trace residue elimination. It acts as a diluent in the application of more viscous PFPE fluids and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text