N-(4-Carboxyphenyl)succinamic acid, 99%, Thermo Scientific Chemicals
N-(4-Carboxyphenyl)succinamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(4-Carboxyphenyl)succinamic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number L17549.06
also known as L17549-06
Price (USD)/ Each
85.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
85.60
Add to cart
N-(4-Carboxyphenyl)succinamic acid, 99%, Thermo Scientific Chemicals
Catalog numberL17549.06
Price (USD)/ Each
85.60
-
Add to cart
Chemical Identifiers
CAS76475-62-2
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only