Thermo Scientific Chemicals

Ethyl 5-methoxyindole-2-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L17596.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 5-methoxyindole-2-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: L17596.06
5 g, Each
Quantity
Catalog number: L17596.06
also known as L17596-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4792-58-9
IUPAC Name
ethyl 5-methoxy-1H-indole-2-carboxylate
Molecular Formula
C12H13NO3
InChI Key
NPIUAXNFAUGNHP-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC2=CC(OC)=CC=C2N1
Appearance (Color)
Dark yellow to yellow to brown
Form
Crystals or powder or crystalline powder or granules
Assay (GC)
≥97.5%
Melting Point (clear melt)
154.0-160.0?C

Description

It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is reactant for preparation of ligands with binding specificity towards the GABA receptor, sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes, anticancer agents,integrase strand-transfer inhibitors (INSTIs),inhibitor of Proliferation of Colon Cancer Cells ,isomeridianin G as GSK-3? inhibitors, HIV-1 integrase inhibitors and inhibitors of mitogen activated protein kinase-activated protein kinase 2 (MK-2).

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text