1-Acetylisatin, 97%, Thermo Scientific Chemicals
1-Acetylisatin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Acetylisatin, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L17597.06
also known as L17597-06
Price (USD)/ Each
70.80
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
70.80
Add to cart
1-Acetylisatin, 97%, Thermo Scientific Chemicals
Catalog numberL17597.06
Price (USD)/ Each
70.80
-
Add to cart
Chemical Identifiers
CAS574-17-4
IUPAC Name1-acetyl-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC10H7NO3
InChI KeyLPGDEHBASRKTDG-UHFFFAOYSA-N
SMILESCC(=O)N1C(=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Melting Point (clear melt)138.0-144.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only